missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromobenzene 97.0+%, TCI America™

Catalog No. p-7126069
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
D016925G 25 g
D0169100G 100 g
D0169500G 500 g
3 options

Catalog No. D016925G

Supplier: TCI America D016925G

Only null left

Chemical Identifiers

CAS 108-36-1
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.91
MDL Number MFCD00000078
InChI Key JSRLURSZEMLAFO-UHFFFAOYSA-N
Synonym m-dibromobenzene, benzene, 1,3-dibromo, benzene, m-dibromo, unii-74ef6kh8tc, 1,3-dibromo-benzene, 74ef6kh8tc, 1,3-dibromo benzene, 3,5-dibromobenzene, 1,3-dibrormobenzene, 1, 3-dibromobenzene
PubChem CID 7927
ChEBI CHEBI:37151
IUPAC Name 1,3-dibromobenzene
SMILES BrC1=CC(Br)=CC=C1

Specifications

Melting Point -7°C
Color Yellow
Boiling Point 219°C
Quantity 25 g
Formula Weight 235.91
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dibromobenzene
RTECSNumber CZ1790000
TSCA Yes