accessibility menu, dialog, popup

UserName

BrBr

CAS RN 108-36-1

IUPAC Name: 1,3-dibromobenzene
Synonyms: 1,3-dibromobenzene M-dibromobenzene
Molecular Weight (g/mol): 235.91
Molecular Formula: C6H4Br2
InChi Key: JSRLURSZEMLAFO-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=CC=C1
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (2)
  • (2)