missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5-nitrobenzene 98.0+%, TCI America™

Catalog No. D53831G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D53831G Supplier TCI America Supplier No. D53831G
Only null left

Chemical Identifiers

CAS 6311-60-0
Molecular Formula C6H3Br2NO2
Molecular Weight (g/mol) 280.903
MDL Number MFCD00092529
InChI Key ZWBHGBWABMAWOV-UHFFFAOYSA-N
Synonym 3,5-Dibromonitrobenzene
PubChem CID 238711
IUPAC Name 1,3-dibromo-5-nitrobenzene
SMILES C1=C(C=C(C=C1Br)Br)[N+](=O)[O-]

Specifications

Melting Point 105°C
Quantity 1 g
Formula Weight 280.90
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibromo-5-nitrobenzene