CAS RN 6311-60-0
IUPAC Name:
1,3-dibromo-5-nitrobenzene
Synonyms:
1,3-dibromo-5-nitrobenzene
Molecular Weight (g/mol):
280.9
Molecular Formula:
C6H3Br2NO2
InChi Key:
ZWBHGBWABMAWOV-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(Br)=CC(Br)=C1
1
–
1
of
1
results
1
1,3-Dibromo-5-nitrobenzene 98.0+%, TCI America™
CAS: 6311-60-0 Molecular Formula: C6H3Br2NO2 Molecular Weight (g/mol): 280.903 MDL Number: MFCD00092529 InChI Key: ZWBHGBWABMAWOV-UHFFFAOYSA-N Synonym: 3,5-Dibromonitrobenzene PubChem CID: 238711 IUPAC Name: 1,3-dibromo-5-nitrobenzene SMILES: C1=C(C=C(C=C1Br)Br)[N+](=O)[O-]