missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5-fluorobenzene 97.0+%, TCI America™

Catalog No. D221110G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D221110G 10 g
D221125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D221110G Supplier TCI America Supplier No. D221110G

Chemical Identifiers

CAS 1435-51-4
Molecular Formula C6H3Br2F
Molecular Weight (g/mol) 253.896
MDL Number MFCD00061119
InChI Key ASWYHZXKFSLNLN-UHFFFAOYSA-N
Synonym 3,5-dibromofluorobenzene, 3,5-dibromo-1-fluorobenzene, 1,3-dibromo-5-fluoro-benzene, benzene, 1,3-dibromo-5-fluoro, pubchem1052, acmc-1c2vf, intermediates-zcf02096, 3,5-bromo-1-fluorophenyl, 1,3dibromo-5-fluorobenzene, 3,5-dibromo-fluoro-benzene
PubChem CID 137003
IUPAC Name 1,3-dibromo-5-fluorobenzene
SMILES C1=C(C=C(C=C1Br)Br)F

Specifications

Color Yellow
Boiling Point 84°C
Quantity 10 g
Formula Weight 253.90
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dibromo-5-fluorobenzene