accessibility menu, dialog, popup

UserName

FBrBr

CAS RN 1435-51-4

IUPAC Name: 1,3-dibromo-5-fluorobenzene
Synonyms: 1,3-dibromo-5-fluorobenzene
Molecular Weight (g/mol): 253.9
Molecular Formula: C6H3Br2F
InChi Key: ASWYHZXKFSLNLN-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC(Br)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)