missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5-fluorobenzene, 98+%

Catalog No. p-7022353
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1507814 25 g
AAA1507822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1507814 Supplier Thermo Scientific Chemicals Supplier No. A1507814
Only null left

CAS: 1435-51-4 | C6H3Br2F | 253.896 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1435-51-4
Molecular Formula C6H3Br2F
Molecular Weight (g/mol) 253.896
MDL Number MFCD00061119
InChI Key ASWYHZXKFSLNLN-UHFFFAOYSA-N
Synonym 3,5-dibromofluorobenzene, 3,5-dibromo-1-fluorobenzene, 1,3-dibromo-5-fluoro-benzene, benzene, 1,3-dibromo-5-fluoro, pubchem1052, acmc-1c2vf, intermediates-zcf02096, 3,5-bromo-1-fluorophenyl, 1,3dibromo-5-fluorobenzene, 3,5-dibromo-fluoro-benzene
PubChem CID 137003
IUPAC Name 1,3-dibromo-5-fluorobenzene
SMILES C1=C(C=C(C=C1Br)Br)F

Specifications

Density 2.018
Boiling Point 204°C to 206°C
Flash Point >110°C (230°F)
Refractive Index 1.577
Quantity 25 g
Beilstein 2353462
Formula Weight 253.91
Percent Purity ≥98%
Chemical Name or Material 1,3-Dibromo-5-fluorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only