missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diamino-2-propanol-N,N,N',N'-tetraacetic Acid 98.0+%, TCI America™
$104.38 - $311.30
Chemical Identifiers
| CAS | 3148-72-9 |
|---|---|
| Molecular Formula | C11H18N2O9 |
| Molecular Weight (g/mol) | 322.27 |
| MDL Number | MFCD00004288 |
| InChI Key | WYMDDFRYORANCC-UHFFFAOYSA-N |
| Synonym | dhpta, dpta, 1,3-diaminopropanol tetraacetic acid, diaminohydroxypropane tetraacetic acid, 1,3-diamino-2-propanol-n,n,n',n'-tetraacetic acid, diaminopropanol tetraacetic acid, unii-949b9zmo7m, 1,3-diamino-2-hydroxypropane-n,n,n',n'-tetraacetic acid, 2-hydroxy-1,3-propylenediaminetetraacetic acid, diaminopropanol tetra acetic acid |
| PubChem CID | 18465 |
| IUPAC Name | 2-[[3-[bis(carboxymethyl)amino]-2-hydroxypropyl]-(carboxymethyl)amino]acetic acid |
| SMILES | C(C(CN(CC(=O)O)CC(=O)O)O)N(CC(=O)O)CC(=O)O |
Chemical Identifiers
| 3148-72-9 | |
| 322.27 | |
| WYMDDFRYORANCC-UHFFFAOYSA-N | |
| 18465 | |
| C(C(CN(CC(=O)O)CC(=O)O)O)N(CC(=O)O)CC(=O)O |
| C11H18N2O9 | |
| MFCD00004288 | |
| dhpta, dpta, 1,3-diaminopropanol tetraacetic acid, diaminohydroxypropane tetraacetic acid, 1,3-diamino-2-propanol-n,n,n',n'-tetraacetic acid, diaminopropanol tetraacetic acid, unii-949b9zmo7m, 1,3-diamino-2-hydroxypropane-n,n,n',n'-tetraacetic acid, 2-hydroxy-1,3-propylenediaminetetraacetic acid, diaminopropanol tetra acetic acid | |
| 2-[[3-[bis(carboxymethyl)amino]-2-hydroxypropyl]-(carboxymethyl)amino]acetic acid |
Specifications
| 3148-72-9 | |
| C11H18N2O9 | |
| 5 g | |
| WYMDDFRYORANCC-UHFFFAOYSA-N | |
| 2-[[3-[bis(carboxymethyl)amino]-2-hydroxypropyl]-(carboxymethyl)amino]acetic acid | |
| 18465 | |
| ≥98.0% (T) | |
| 1,3-Diamino-2-propanol-N,N,N′,N′-tetraacetic Acid |
| White | |
| MFCD00004288 | |
| dhpta, dpta, 1,3-diaminopropanol tetraacetic acid, diaminohydroxypropane tetraacetic acid, 1,3-diamino-2-propanol-n,n,n',n'-tetraacetic acid, diaminopropanol tetraacetic acid, unii-949b9zmo7m, 1,3-diamino-2-hydroxypropane-n,n,n',n'-tetraacetic acid, 2-hydroxy-1,3-propylenediaminetetraacetic acid, diaminopropanol tetra acetic acid | |
| C(C(CN(CC(=O)O)CC(=O)O)O)N(CC(=O)O)CC(=O)O | |
| 322.27 | |
| 322.27 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-2681
RTECSNumber : AI2930000
TSCA : Yes