CAS RN 3148-72-9
IUPAC Name:
2-({3-[bis(carboxymethyl)amino]-2-hydroxypropyl}(carboxymethyl)amino)acetic acid
Synonyms:
({3-[bis(carboxymethyl)amino]-2-hydroxypropyl}(carboxymethyl)amino)acetic acid
Molecular Weight (g/mol):
322.27
Molecular Formula:
C11H18N2O9
InChi Key:
WYMDDFRYORANCC-UHFFFAOYSA-N
SMILES:
OC(CN(CC(O)=O)CC(O)=O)CN(CC(O)=O)CC(O)=O
1
–
1
of
1
results
1
1,3-Diamino-2-propanol-N,N,N',N'-tetraacetic Acid 98.0+%, TCI America™
CAS: 3148-72-9 Molecular Formula: C11H18N2O9 Molecular Weight (g/mol): 322.27 MDL Number: MFCD00004288 InChI Key: WYMDDFRYORANCC-UHFFFAOYSA-N Synonym: dhpta,dpta,1,3-diaminopropanol tetraacetic acid,diaminohydroxypropane tetraacetic acid,1,3-diamino-2-propanol-n,n,n',n'-tetraacetic acid,diaminopropanol tetraacetic acid,unii-949b9zmo7m,1,3-diamino-2-hydroxypropane-n,n,n',n'-tetraacetic acid,2-hydroxy-1,3-propylenediaminetetraacetic acid,diaminopropanol tetra acetic acid PubChem CID: 18465 IUPAC Name: 2-[[3-[bis(carboxymethyl)amino]-2-hydroxypropyl]-(carboxymethyl)amino]acetic acid SMILES: C(C(CN(CC(=O)O)CC(=O)O)O)N(CC(=O)O)CC(=O)O