missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diamino-2-propanol-N,N,N',N'-tetraacetic Acid 98.0+%, TCI America™
Supplier: TCI America D133025G
Specifications
| 1,3-Diamino-2-propanol-N,N,N′,N′-tetraacetic Acid | |
| White | |
| MFCD00004288 | |
| dhpta, dpta, 1,3-diaminopropanol tetraacetic acid, diaminohydroxypropane tetraacetic acid, 1,3-diamino-2-propanol-n,n,n',n'-tetraacetic acid, diaminopropanol tetraacetic acid, unii-949b9zmo7m, 1,3-diamino-2-hydroxypropane-n,n,n',n'-tetraacetic acid, 2-hydroxy-1,3-propylenediaminetetraacetic acid, diaminopropanol tetra acetic acid | |
| C(C(CN(CC(=O)O)CC(=O)O)O)N(CC(=O)O)CC(=O)O | |
| 322.27 | |
| 322.27 | |
| Crystalline Powder |
| 3148-72-9 | |
| C11H18N2O9 | |
| 25 g | |
| WYMDDFRYORANCC-UHFFFAOYSA-N | |
| 2-[[3-[bis(carboxymethyl)amino]-2-hydroxypropyl]-(carboxymethyl)amino]acetic acid | |
| 18465 | |
| ≥98.0% (T) |
Chemical Identifiers
| 3148-72-9 | |
| 322.27 | |
| WYMDDFRYORANCC-UHFFFAOYSA-N | |
| 18465 | |
| C(C(CN(CC(=O)O)CC(=O)O)O)N(CC(=O)O)CC(=O)O |
| C11H18N2O9 | |
| MFCD00004288 | |
| dhpta, dpta, 1,3-diaminopropanol tetraacetic acid, diaminohydroxypropane tetraacetic acid, 1,3-diamino-2-propanol-n,n,n',n'-tetraacetic acid, diaminopropanol tetraacetic acid, unii-949b9zmo7m, 1,3-diamino-2-hydroxypropane-n,n,n',n'-tetraacetic acid, 2-hydroxy-1,3-propylenediaminetetraacetic acid, diaminopropanol tetra acetic acid | |
| 2-[[3-[bis(carboxymethyl)amino]-2-hydroxypropyl]-(carboxymethyl)amino]acetic acid |
Safety and Handling
EINECSNumber : (2)-2681
RTECSNumber : AI2930000
TSCA : Yes