missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-tert-butyl-2-methoxybenzene, 99%

Catalog No. aah3748709
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3748709 10 g
AAH3748718 50 g
AAH3748730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3748709 Supplier Thermo Scientific Chemicals Supplier No. H3748709
Only null left

CAS: 1516-95-6 | C15H24O | 220.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1516-95-6
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.36
MDL Number MFCD20483412
InChI Key OGIJRUWUWSZKAV-UHFFFAOYSA-N
Synonym 1,3-di-tert-butyl-2-methoxybenzene, benzene, 1,3-bis 1,1-dimethylethyl-2-methoxy, 2,6-di-tert-butylanisole
PubChem CID 12794694
IUPAC Name 1,3-ditert-butyl-2-methoxybenzene
SMILES COC1=C(C=CC=C1C(C)(C)C)C(C)(C)C

Specifications

Color Colorless to Yellow
Boiling Point 124°C (9 mmHg)
Quantity 10 g
Formula Weight 220.35
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,3-Di-tert-butyl-2-methoxybenzene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only