missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-O-benzyl-2-O-lauroylglycerol 85.0+%, TCI America™

Catalog No. D35241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35241G 1 g
D35245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35241G Supplier TCI America Supplier No. D35241G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 519177-07-2
Molecular Formula C29H42O4
Molecular Weight (g/mol) 454.65
MDL Number MFCD09038522
InChI Key HMRAWECRZFFMJY-UHFFFAOYSA-N
Synonym Glycerol 1,3-Dibenzyl Ether 2-Laurate, 1,3-Bis(benzyloxy)isopropyl Dodecanoate
PubChem CID 44629829
IUPAC Name 1,3-bis(benzyloxy)propan-2-yl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC(COCC1=CC=CC=C1)COCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 454.65
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Di-O-benzyl-2-O-lauroylglycerol