CAS RN 519177-07-2
CAS RN 519177-07-2
IUPAC Name:
1,3-bis(benzyloxy)propan-2-yl dodecanoate
Synonyms:
1,3-bis(benzyloxy)propan-2-yl dodecanoate
Molecular Weight (g/mol):
454.65
Molecular Formula:
C29H42O4
InChi Key:
HMRAWECRZFFMJY-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC(=O)OC(COCC1=CC=CC=C1)COCC1=CC=CC=C1
1
–
1
of
1
results
1
1,3-Di-O-benzyl-2-O-lauroylglycerol 85.0+%, TCI America™
CAS: 519177-07-2 Molecular Formula: C29H42O4 Molecular Weight (g/mol): 454.65 MDL Number: MFCD09038522 InChI Key: HMRAWECRZFFMJY-UHFFFAOYSA-N Synonym: Glycerol 1,3-Dibenzyl Ether 2-Laurate, 1,3-Bis(benzyloxy)isopropyl Dodecanoate PubChem CID: 44629829 IUPAC Name: 1,3-bis(benzyloxy)propan-2-yl dodecanoate SMILES: CCCCCCCCCCCC(=O)OC(COCC1=CC=CC=C1)COCC1=CC=CC=C1