missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Bromopropyl)pyrrole 97.0+%, TCI America™

Catalog No. p-7119165
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16231G 1 g
B16235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16231G Supplier TCI America Supplier No. B16231G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100779-91-7
Molecular Formula C7H10BrN
Molecular Weight (g/mol) 188.07
MDL Number MFCD00191324
InChI Key IXIXPLRTYIMRMC-UHFFFAOYSA-N
PubChem CID 557119
IUPAC Name 1-(3-bromopropyl)-1H-pyrrole
SMILES BrCCCN1C=CC=C1

Specifications

Color Yellow
Boiling Point 78°C
Quantity 1 g
Formula Weight 188.07
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(3-Bromopropyl)pyrrole
TSCA No
Recommended Storage Refrigerator