accessibility menu, dialog, popup

UserName

CAS RN 100779-91-7

NBr

CAS RN 100779-91-7

IUPAC Name: 1-(3-bromopropyl)-1H-pyrrole
Synonyms: 1-(3-bromopropyl)pyrrole
Molecular Weight (g/mol): 188.07
Molecular Formula: C7H10BrN
InChi Key: IXIXPLRTYIMRMC-UHFFFAOYSA-N
SMILES: BrCCCN1C=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results