missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis(4-fluorophenyl)urea 98.0+%, TCI America™
Supplier: TCI America B45175G
Specifications
| 1,3-Bis(4-fluorophenyl)urea | |
| 267°C | |
| C13H10F2N2O | |
| 5 g | |
| VEURDHASKSZBOL-UHFFFAOYSA-N | |
| 1,3-bis(4-fluorophenyl)urea | |
| 302982 | |
| ≥98.0% (HPLC,N) |
| 370-22-9 | |
| White | |
| MFCD00017925 | |
| 1,3-bis 4-fluorophenyl urea, 1,3-di 4-fluorophenyl urea, n,n'-bis 4-fluorophenyl urea, n,n'-bis 4-fluorophenyl urea #, urea, n,n'-bis 4-fluorophenyl, n-4-fluorophenyl 4-fluorophenyl amino carboxamide | |
| FC1=CC=C(NC(=O)NC2=CC=C(F)C=C2)C=C1 | |
| 248.23 | |
| 248.23 | |
| Crystalline Powder |
Chemical Identifiers
| 370-22-9 | |
| 248.23 | |
| VEURDHASKSZBOL-UHFFFAOYSA-N | |
| 302982 | |
| FC1=CC=C(NC(=O)NC2=CC=C(F)C=C2)C=C1 |
| C13H10F2N2O | |
| MFCD00017925 | |
| 1,3-bis 4-fluorophenyl urea, 1,3-di 4-fluorophenyl urea, n,n'-bis 4-fluorophenyl urea, n,n'-bis 4-fluorophenyl urea #, urea, n,n'-bis 4-fluorophenyl, n-4-fluorophenyl 4-fluorophenyl amino carboxamide | |
| 1,3-bis(4-fluorophenyl)urea |
Safety and Handling
TSCA : No