accessibility menu, dialog, popup

UserName

CAS RN 370-22-9

IUPAC Name: 1,3-bis(4-fluorophenyl)urea
Synonyms: 1,3-bis(4-fluorophenyl)urea
Molecular Weight (g/mol): 248.23
Molecular Formula: C13H10F2N2O
InChi Key: VEURDHASKSZBOL-UHFFFAOYSA-N
SMILES: FC1=CC=C(NC(=O)NC2=CC=C(F)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results