missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(4-fluorophenyl)urea 98.0+%, TCI America™

Catalog No. p-7123590
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45171G 1 g
B45175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45171G Supplier TCI America Supplier No. B45171G

Chemical Identifiers

CAS 370-22-9
Molecular Formula C13H10F2N2O
Molecular Weight (g/mol) 248.23
MDL Number MFCD00017925
InChI Key VEURDHASKSZBOL-UHFFFAOYSA-N
Synonym 1,3-bis 4-fluorophenyl urea, 1,3-di 4-fluorophenyl urea, n,n'-bis 4-fluorophenyl urea, n,n'-bis 4-fluorophenyl urea #, urea, n,n'-bis 4-fluorophenyl, n-4-fluorophenyl 4-fluorophenyl amino carboxamide
PubChem CID 302982
IUPAC Name 1,3-bis(4-fluorophenyl)urea
SMILES FC1=CC=C(NC(=O)NC2=CC=C(F)C=C2)C=C1

Specifications

Melting Point 267°C
Color White
Quantity 1 g
Formula Weight 248.23
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(4-fluorophenyl)urea