missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(4-chlorophenyl)urea 98.0+%, TCI America™

Catalog No. B45291G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45291G 1 g
B45295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45291G Supplier TCI America Supplier No. B45291G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1219-99-4
Molecular Formula C13H10Cl2N2O
Molecular Weight (g/mol) 281.14
MDL Number MFCD00018541
InChI Key ZNQCSLYENQIUMJ-UHFFFAOYSA-N
PubChem CID 94996
IUPAC Name 1,3-bis(4-chlorophenyl)urea
SMILES ClC1=CC=C(NC(=O)NC2=CC=C(Cl)C=C2)C=C1

Specifications

Melting Point 306°C
Color White
Quantity 1 g
Formula Weight 281.14
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(4-chlorophenyl)urea