missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(3-aminophenoxy)benzene 98.0+%, TCI America™

Catalog No. B137025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B137025G 25 g
B13705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B137025G Supplier TCI America Supplier No. B137025G
Only null left

Chemical Identifiers

CAS 10526-07-5
Molecular Formula C18H16N2O2
Molecular Weight (g/mol) 292.338
MDL Number MFCD00043718
InChI Key DKKYOQYISDAQER-UHFFFAOYSA-N
Synonym Resorcinol Bis(3-aminophenyl) Ether
PubChem CID 66342
IUPAC Name 3-[3-(3-aminophenoxy)phenoxy]aniline
SMILES C1=CC(=CC(=C1)OC2=CC(=CC=C2)OC3=CC=CC(=C3)N)N

Specifications

Melting Point 108°C
Quantity 25 g
Formula Weight 292.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(3-aminophenoxy)benzene
EINECSNumber (3)-3948
TSCA Yes