accessibility menu, dialog, popup

UserName

NH2OONH2

CAS RN 10526-07-5

IUPAC Name: 3-[3-(3-aminophenoxy)phenoxy]aniline
Synonyms: 3-[3-(3-aminophenoxy)phenoxy]aniline
Molecular Weight (g/mol): 292.34
Molecular Formula: C18H16N2O2
InChi Key: DKKYOQYISDAQER-UHFFFAOYSA-N
SMILES: NC1=CC(OC2=CC(OC3=CC(N)=CC=C3)=CC=C2)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results