missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea 98.0+%, TCI America™

Catalog No. B3452200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3452200MG 200 mg
B34521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3452200MG Supplier TCI America Supplier No. B3452200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1060-92-0
Molecular Formula C17H8F12N2S
Molecular Weight (g/mol) 500.306
InChI Key RWXWQJYJWJNJNW-UHFFFAOYSA-N
PubChem CID 2803772
IUPAC Name 1,3-bis[3,5-bis(trifluoromethyl)phenyl]thiourea
SMILES C1=C(C=C(C=C1C(F)(F)F)NC(=S)NC2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 200 mg
Formula Weight 500.31
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea