accessibility menu, dialog, popup

UserName

CAS RN 1060-92-0

IUPAC Name: 1,3-bis[3,5-bis(trifluoromethyl)phenyl]thiourea
Synonyms: 1,3-bis[3,5-bis(trifluoromethyl)phenyl]thiourea
Molecular Weight (g/mol): 500.31
Molecular Formula: C17H8F12N2S
InChi Key: RWXWQJYJWJNJNW-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(NC(=S)NC2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)