missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Alpha Vector | BISBISTRIFLUOROMETHYLPHENYLTHIOUREA |
| Melting Point | 150°C to 157°C |
| Color | Brown to White |
| Infrared Spectrum | Conforms |
| Packaging | glass bottle |
| Quantity | 5 g |
| Formula Weight | 500.3 |
| Physical Form | Crsytalline Powder or Crystals |
| Chemical Name or Material | 1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea |
Safety and Handling
| Hazard Category | GHS07: Harmful |
| ShelfLife | 5 years |
| storageNote1 | normal conditions |
RUO – Research Use Only