missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea

Catalog No. AC465880010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465880010 1 g
AC465880050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465880010 Supplier Thermo Scientific Chemicals Supplier No. 465880010
Only null left

CAS: 1060-92-0 | C17H8F12N2S | 500.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1060-92-0
Molecular Formula C17H8F12N2S
Molecular Weight (g/mol) 500.31
MDL Number MFCD00829878
InChI Key RWXWQJYJWJNJNW-UHFFFAOYSA-N
Synonym Schreiner's Thiourea Catalyst
IUPAC Name 1,3-bis[3,5-bis(trifluoromethyl)phenyl]thiourea
SMILES FC(F)(F)C1=CC(=CC(NC(=S)NC2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)=C1)C(F)(F)F

Specifications

Alpha Vector BISBISTRIFLUOROMETHYLPHENYLTHIOUREA
Melting Point 150°C to 157°C
Color Brown to White
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 1 g
Formula Weight 500.3
Physical Form Crsytalline Powder or Crystals
Chemical Name or Material 1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea
Hazard Category GHS07: Harmful
ShelfLife 5 years
storageNote1 normal conditions

RUO – Research Use Only