missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis(3,3,3-trifluoropropyl)-1,1,3,3-tetramethyldisilazane 94.0+%, TCI America™
Supplier: TCI America B331925G
Specifications
| 1,3-Bis(3,3,3-trifluoropropyl)-1,1,3,3-tetramethyldisilazane | |
| Yellow | |
| C10H21F6NSi2 | |
| 25 g | |
| C[Si](C)(CCC(F)(F)F)N[Si](C)(C)CCC(F)(F)F | |
| 325.445 | |
| 325.45 | |
| Liquid |
| 39482-87-6 | |
| 119°C | |
| MFCD00153070 | |
| ITRFWRDOAWGZFV-UHFFFAOYSA-N | |
| 3-[[[dimethyl(3,3,3-trifluoropropyl)silyl]amino]-dimethylsilyl]-1,1,1-trifluoropropane | |
| 170213 | |
| ≥94.0% (GC) |
Chemical Identifiers
| 39482-87-6 | |
| 325.445 | |
| ITRFWRDOAWGZFV-UHFFFAOYSA-N | |
| 3-[[[dimethyl(3,3,3-trifluoropropyl)silyl]amino]-dimethylsilyl]-1,1,1-trifluoropropane |
| C10H21F6NSi2 | |
| MFCD00153070 | |
| 170213 | |
| C[Si](C)(CCC(F)(F)F)N[Si](C)(C)CCC(F)(F)F |
Safety and Handling
TSCA : Yes