missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(3,3,3-trifluoropropyl)-1,1,3,3-tetramethyldisilazane 94.0+%, TCI America™

Catalog No. p-7122871
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33195G 5 g
B331925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33195G Supplier TCI America Supplier No. B33195G
Only null left

Chemical Identifiers

CAS 39482-87-6
Molecular Formula C10H21F6NSi2
Molecular Weight (g/mol) 325.445
MDL Number MFCD00153070
InChI Key ITRFWRDOAWGZFV-UHFFFAOYSA-N
PubChem CID 170213
IUPAC Name 3-[[[dimethyl(3,3,3-trifluoropropyl)silyl]amino]-dimethylsilyl]-1,1,1-trifluoropropane
SMILES C[Si](C)(CCC(F)(F)F)N[Si](C)(C)CCC(F)(F)F

Specifications

Color Yellow
Boiling Point 119°C
Quantity 5 g
Formula Weight 325.45
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Bis(3,3,3-trifluoropropyl)-1,1,3,3-tetramethyldisilazane