accessibility menu, dialog, popup

UserName

CAS RN 39482-87-6

CH3SiH3CFFFNHSiCH3H3CFFF

CAS RN 39482-87-6

IUPAC Name: bis[dimethyl(3,3,3-trifluoropropyl)silyl]amine
Synonyms: bis[dimethyl(3,3,3-trifluoropropyl)silyl]amine
Molecular Weight (g/mol): 325.45
Molecular Formula: C10H21F6NSi2
InChi Key: ITRFWRDOAWGZFV-UHFFFAOYSA-N
SMILES: C[Si](C)(CCC(F)(F)F)N[Si](C)(C)CCC(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results