missing translation for 'onlineSavingsMsg'
Learn More

[1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene]triphenylphosphine Nickel(II) Dichloride 98.0+%, TCI America™

Catalog No. B3235200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3235200MG 200 mg
B32351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3235200MG Supplier TCI America Supplier No. B3235200MG

Chemical Identifiers

CAS 903592-98-3
Molecular Formula C45H51Cl2N2NiP
Molecular Weight (g/mol) 780.484
InChI Key IGLKEXXECLLIAD-UHFFFAOYSA-L
PubChem CID 56923621
IUPAC Name [1,3-bis[2,6-di(propan-2-yl)phenyl]imidazol-2-ylidene]-triphenyl-$l^{5}-phosphane;dichloronickel
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2C=CN(C2=P(C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5)C6=C(C=CC=C6C(C)C)C(C)C.Cl[Ni]Cl

Specifications

Color Red-Yellow
Quantity 200 mg
Formula Weight 780.48
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene]triphenylphosphine Nickel(II) Dichloride