accessibility menu, dialog, popup

UserName

CH3CH3H3CCH3NNPCH3H3CCH3CH3ClNiCl(?)(?)

CAS RN 903592-98-3

IUPAC Name: 1,3-bis[2,6-bis(propan-2-yl)phenyl]-2-(triphenyl-λ⁵-phosphanylidene)-2,3-dihydro-1H-imidazole; dichloronickel
Synonyms: 1,3-bis(2,6-diisopropylphenyl)-2-(triphenyl-λ⁵-phosphanylidene)imidazole; nickel chloride
Molecular Weight (g/mol): 780.48
Molecular Formula: C45H51Cl2N2NiP
InChi Key: IGLKEXXECLLIAD-UHFFFAOYSA-L
SMILES: Cl[Ni]Cl.CC(C)C1=CC=CC(C(C)C)=C1N1C=CN(C1=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC=C1C(C)C)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results