missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene 98.0+%, TCI America™

Catalog No. B266425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B266425G 25 g
B2664250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B266425G Supplier TCI America Supplier No. B266425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13595-25-0
Molecular Formula C24H26O2
Molecular Weight (g/mol) 346.47
InChI Key PVFQHGDIOXNKIC-UHFFFAOYSA-N
Synonym Bisphenol M, 4,4′C-(1,3-Phenylenediisopropylidene)bisphenol
PubChem CID 3292100
IUPAC Name 4-[2-[3-[2-(4-hydroxyphenyl)propan-2-yl]phenyl]propan-2-yl]phenol
SMILES CC(C)(C1=CC=C(C=C1)O)C2=CC(=CC=C2)C(C)(C)C3=CC=C(C=C3)O

Specifications

Melting Point 138°C
Color White
Quantity 25 g
Formula Weight 346.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene
EINECSNumber (4)-1582
TSCA Yes