accessibility menu, dialog, popup

UserName

CAS RN 13595-25-0

IUPAC Name: 4-(1-{3-[2-(4-hydroxyphenyl)propyl]phenyl}propan-2-yl)phenol
Synonyms: 4-(1-{3-[2-(4-hydroxyphenyl)propyl]phenyl}propan-2-yl)phenol
Molecular Weight (g/mol): 346.47
Molecular Formula: C24H26O2
InChi Key: KOWHWSRFLBFYRR-UHFFFAOYNA-N
SMILES: CC(CC1=CC(CC(C)C2=CC=C(O)C=C2)=CC=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results