missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(4-bromophenyl)benzene 98.0+%, TCI America™

Catalog No. p-7137334
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26441G 1 g
T26445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26441G Supplier TCI America Supplier No. T26441G
Only null left

Chemical Identifiers

CAS 7511-49-1
Molecular Formula C24H15Br3
Molecular Weight (g/mol) 543.096
MDL Number MFCD00362911
InChI Key HJQRITCAXSBOPC-UHFFFAOYSA-N
Synonym 1,3,5-tris 4-bromophenyl benzene, 4,4-dibromo-5'-4-bromophenyl-1,1':3',1-terphenyl, 1,1':3',1-terphenyl, 4,4-dibromo-5'-4-bromophenyl, 4'-bromo-3,5-bis 4-bromophenyl-1,1'-biphenyl, acmc-209ox3, ksc496e7d, 1,3,5-tris p-bromophenyl benzene, 1,3,5-tri 4-bromophenyl benzene, 4,4-dibromo-5'-4-bromophenyl-1,1'
PubChem CID 232761
IUPAC Name 1,3,5-tris(4-bromophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC(=CC(=C2)C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br)Br

Specifications

Melting Point 263°C
Quantity 1 g
Formula Weight 543.10
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris(4-bromophenyl)benzene