missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-1,3,5-triazinane-2,4,6-trione 98.0+%, TCI America™

Catalog No. T3653500G
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3653500G 500 g
T3653100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3653500G Supplier TCI America Supplier No. T3653500G

Chemical Identifiers

CAS 27676-62-6
Molecular Formula C48H69N3O6
Molecular Weight (g/mol) 784.10
MDL Number MFCD00134700
InChI Key VNQNXQYZMPJLQX-UHFFFAOYSA-N
Synonym Tris(3,5-di-tert-butyl-4-hydroxybenzyl) Isocyanurate, Isocyanuric Acid Tris(3,5-di-tert-butyl-4-hydroxybenzyl) Ester
PubChem CID 93115
IUPAC Name tris[(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]-1,3,5-triazinane-2,4,6-trione
SMILES CC(C)(C)C1=CC(CN2C(=O)N(CC3=CC(=C(O)C(=C3)C(C)(C)C)C(C)(C)C)C(=O)N(CC3=CC(=C(O)C(=C3)C(C)(C)C)C(C)(C)C)C2=O)=CC(=C1O)C(C)(C)C

Specifications

Melting Point 222°C
Color White-Yellow
Quantity 500 g
Formula Weight 784.10
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-1,3,5-triazinane-2,4,6-trione
EINECSNumber (5)-1073
TSCA Yes