accessibility menu, dialog, popup

UserName

CH3H3CCH3NONOHCH3H3CCH3CH3CH3H3CONOHH3CH3CCH3CH3H3CCH3OHOH3CH3CCH3

CAS RN 27676-62-6

IUPAC Name: tris[(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]-1,3,5-triazinane-2,4,6-trione
Synonyms: tris[(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]-1,3,5-triazinane-2,4,6-trione
Molecular Weight (g/mol): 784.1
Molecular Formula: C48H69N3O6
InChi Key: VNQNXQYZMPJLQX-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(CN2C(=O)N(CC3=CC(=C(O)C(=C3)C(C)(C)C)C(C)(C)C)C(=O)N(CC3=CC(=C(O)C(=C3)C(C)(C)C)C(C)(C)C)C2=O)=CC(=C1O)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results