missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Triisopropylbenzene 95.0+%, TCI America™
Supplier: TCI America T0458500ML
Specifications
| 1,3,5-Triisopropylbenzene | |
| Colorless | |
| C15H24 | |
| 500 mL | |
| VUMCUSHVMYIRMB-UHFFFAOYSA-N | |
| 1,3,5-tri(propan-2-yl)benzene | |
| 12860 | |
| ≥95.0% (GC) |
| 717-74-8 | |
| 235°C | |
| MFCD00008890 | |
| 1,3,5-triisopropylbenzene, 2,4,6-triisopropylbenzene, benzene, 1,3,5-tris 1-methylethyl, 1,3,5-tris propan-2-yl benzene, unii-fr9y346wpb, benzene, 1,3,5-triisopropyl, fr9y346wpb, benzene, tris 1-methylethyl, pubchem13720, 1,5-triisopropylbenzene | |
| CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C | |
| 204.357 | |
| 204.36 | |
| Liquid |
Chemical Identifiers
| 717-74-8 | |
| 204.357 | |
| VUMCUSHVMYIRMB-UHFFFAOYSA-N | |
| 12860 | |
| CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C |
| C15H24 | |
| MFCD00008890 | |
| 1,3,5-triisopropylbenzene, 2,4,6-triisopropylbenzene, benzene, 1,3,5-tris 1-methylethyl, 1,3,5-tris propan-2-yl benzene, unii-fr9y346wpb, benzene, 1,3,5-triisopropyl, fr9y346wpb, benzene, tris 1-methylethyl, pubchem13720, 1,5-triisopropylbenzene | |
| 1,3,5-tri(propan-2-yl)benzene |
Safety and Handling
EINECSNumber : (3)-0022&(3)-3427
TSCA : Yes