missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triisopropylbenzene 95.0+%, TCI America™

Catalog No. T045825ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T045825ML 25 mL
T0458100ML 100 mL
T0458500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T045825ML Supplier TCI America Supplier No. T045825ML

Chemical Identifiers

CAS 717-74-8
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
MDL Number MFCD00008890
InChI Key VUMCUSHVMYIRMB-UHFFFAOYSA-N
Synonym 1,3,5-triisopropylbenzene, 2,4,6-triisopropylbenzene, benzene, 1,3,5-tris 1-methylethyl, 1,3,5-tris propan-2-yl benzene, unii-fr9y346wpb, benzene, 1,3,5-triisopropyl, fr9y346wpb, benzene, tris 1-methylethyl, pubchem13720, 1,5-triisopropylbenzene
PubChem CID 12860
IUPAC Name 1,3,5-tri(propan-2-yl)benzene
SMILES CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C

Specifications

Color Colorless
Boiling Point 235°C
Quantity 25 mL
Formula Weight 204.36
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5-Triisopropylbenzene
EINECSNumber (3)-0022&(3)-3427
TSCA Yes