missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triisopropylbenzene, 95%

Catalog No. p-205590
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC171220250 25 mL Glass bottle
AC171221000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC171220250 Supplier Thermo Scientific Chemicals Supplier No. 171220250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 717-74-8 | C15H24 | 204.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 717-74-8
Molecular Formula C15H24
Molecular Weight (g/mol) 204.35
MDL Number MFCD00008890
InChI Key VUMCUSHVMYIRMB-UHFFFAOYSA-N
Synonym 1,3,5-triisopropylbenzene, 2,4,6-triisopropylbenzene, benzene, 1,3,5-tris 1-methylethyl, 1,3,5-tris propan-2-yl benzene, unii-fr9y346wpb, benzene, 1,3,5-triisopropyl, fr9y346wpb, benzene, tris 1-methylethyl, pubchem13720, 1,5-triisopropylbenzene
PubChem CID 12860
IUPAC Name 1,3,5-tri(propan-2-yl)benzene
SMILES CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 0.8400g/mL
Boiling Point 232°C to 236°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4874 to 1.4894
Linear Formula C6H3[CH(CH3)2]3
Quantity 25 mL
Beilstein 05, 458
Specific Gravity 0.84
Formula Weight 204.35
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1, 3, 5-Triisopropylbenzene, 97%
Show More Show Less
EINECSNumber 211-941-3
TSCA TSCA

RUO – Research Use Only