missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triisopropylbenzene, 95%

CAS: 717-74-8 | C15H24 | 204.357 g/mol

Supplier:  Thermo Scientific Chemicals A134530B

 View more versions of this product

Catalog No. AAA134530B


Only null left
Explore available promotions

Description

Description

1,3,5-Triisopropylbenzene is used as a fuel and fuel additive. It is also used in lubricants and lubricant additives. Further, it is used in the preparation of 2,4,6-triisopropyl-benzenesulfonyl chloride. It is also employed as a swelling agent during the synthesis of mesoporous silica with two-dimensional hexagonal pores. In addition to this, it is used as a micelle expander in the synthesis of magnetic mesoporous silica nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Triisopropylbenzene is used as a fuel and fuel additive. It is also used in lubricants and lubricant additives. Further, it is used in the preparation of 2,4,6-triisopropyl-benzenesulfonyl chloride. It is also employed as a swelling agent during the synthesis of mesoporous silica with two-dimensional hexagonal pores. In addition to this, it is used as a micelle expander in the synthesis of magnetic mesoporous silica nanoparticles.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

1,3,5-Triisopropylbenzene
-7°C
235°C to 237°C
C15H24
MFCD00008890
1862750
Immiscible with water.
CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C
204.357
204.36
717-74-8
0.854
86°C (186°F)
1.488
1000 g
1,3,5-triisopropylbenzene, 2,4,6-triisopropylbenzene, benzene, 1,3,5-tris 1-methylethyl, 1,3,5-tris propan-2-yl benzene, unii-fr9y346wpb, benzene, 1,3,5-triisopropyl, fr9y346wpb, benzene, tris 1-methylethyl, pubchem13720, 1,5-triisopropylbenzene
VUMCUSHVMYIRMB-UHFFFAOYSA-N
1,3,5-tri(propan-2-yl)benzene
12860
95%

Chemical Identifiers

717-74-8
204.357
VUMCUSHVMYIRMB-UHFFFAOYSA-N
12860
CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C
C15H24
MFCD00008890
1,3,5-triisopropylbenzene, 2,4,6-triisopropylbenzene, benzene, 1,3,5-tris 1-methylethyl, 1,3,5-tris propan-2-yl benzene, unii-fr9y346wpb, benzene, 1,3,5-triisopropyl, fr9y346wpb, benzene, tris 1-methylethyl, pubchem13720, 1,5-triisopropylbenzene
1,3,5-tri(propan-2-yl)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P361-P363-P370+P378q-P501c

H227-H311-H315-H319

EINECSNumber : 211-941-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only