missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triformylbenzene, 98%

Catalog No. AC446550010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446550010 1 g
AC446550050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446550010 Supplier Thermo Scientific Chemicals Supplier No. 446550010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3163-76-6 | C9H6O3 | 162.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3163-76-6
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00182470
InChI Key AEKQNAANFVOBCU-UHFFFAOYSA-N
Synonym 1,3,5-benzenetricarboxaldehyde, 1,3,5-triformylbenzene, trimesaldehyde, 1,3,5-benzenetricarbaldehyde, benzene-1,3,5-tricarboxaldehyde, trimesic formaldehyde, 1,3,5-triformyl benzene, bzoh31, aekqnaanfvobcu-uhfffaoysa
PubChem CID 2747968
IUPAC Name benzene-1,3,5-tricarbaldehyde
SMILES O=CC1=CC(C=O)=CC(C=O)=C1

Specifications

Melting Point 156.0°C to 158.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 162.14
Percent Purity 98%
Chemical Name or Material 1, 3, 5-Triformylbenzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only