missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%

Catalog No. AAH3311603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3311603 1 g
AAH3311606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3311603 Supplier Thermo Scientific Chemicals Supplier No. H3311603

CAS: 84322-56-5 | C6F3I3 | 509.775 g/mol

1,3,5-Trifluoro-2,4,6-triiodobenzene is useful for biological applications. Further, it is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trifluoro-2,4,6-triiodobenzene is useful for biological applications. Further, it is used as an intermediate in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 84322-56-5
Molecular Formula C6F3I3
Molecular Weight (g/mol) 509.775
MDL Number MFCD06248899
InChI Key NTAZOPPTLZSXQH-UHFFFAOYSA-N
Synonym 1,3,5-trifluoro-2,4,6-triiodo-benzene, 1,3,5-trilfluoro-2,4,6-triiodobenzene, benzene,1,3,5-trifluoro-2,4,6-triiodo, pubchem10637, 1,3,5-trifluoro-2,4, 1,3,5-triiodotrifluorobenzene, 1,3,5-trifluoro-2,4,6-triiodob, benzene, 1,3,5-trifluoro-2,4,6-triiodo
PubChem CID 3852430
IUPAC Name 1,3,5-trifluoro-2,4,6-triiodobenzene
SMILES C1(=C(C(=C(C(=C1I)F)I)F)I)F

Specifications

Melting Point 152°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 509.77
Percent Purity 97%
Chemical Name or Material 1,3,5-Trifluoro-2,4,6-triiodobenzene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only