accessibility menu, dialog, popup

UserName

IFIFIF

CAS RN 84322-56-5

IUPAC Name: 1,3,5-trifluoro-2,4,6-triiodobenzene
Synonyms: 1,3,5-trifluoro-2,4,6-triiodobenzene
Molecular Weight (g/mol): 509.78
Molecular Formula: C6F3I3
InChi Key: NTAZOPPTLZSXQH-UHFFFAOYSA-N
SMILES: FC1=C(I)C(F)=C(I)C(F)=C1I
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results