missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%

Catalog No. p-7077393
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452690010 1 g
AC452690050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452690010 Supplier Thermo Scientific Chemicals Supplier No. 452690010

CAS: 84322-56-5 | C6F3I3 | 509.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84322-56-5
Molecular Formula C6F3I3
Molecular Weight (g/mol) 509.78
InChI Key NTAZOPPTLZSXQH-UHFFFAOYSA-N
Synonym 1,3,5-trifluoro-2,4,6-triiodo-benzene, 1,3,5-trilfluoro-2,4,6-triiodobenzene, benzene,1,3,5-trifluoro-2,4,6-triiodo, pubchem10637, 1,3,5-trifluoro-2,4, 1,3,5-triiodotrifluorobenzene, 1,3,5-trifluoro-2,4,6-triiodob, benzene, 1,3,5-trifluoro-2,4,6-triiodo
PubChem CID 3852430
IUPAC Name 1,3,5-trifluoro-2,4,6-triiodobenzene
SMILES C1(=C(C(=C(C(=C1I)F)I)F)I)F

Specifications

Boiling Point 154.0°C to 156.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 509.78
Percent Purity 97%
Chemical Name or Material 1,3,5-Trifluoro-2,4,6-triiodobenzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only