missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri-tert-butylbenzene 98.0+%, TCI America™

Catalog No. T16335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16335G 5 g
T163325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16335G Supplier TCI America Supplier No. T16335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1460-02-2
Molecular Formula C18H30
Molecular Weight (g/mol) 246.438
MDL Number MFCD00008831
InChI Key GUFMBISUSZUUCB-UHFFFAOYSA-N
Synonym 1,3,5-tri-tert-butylbenzene, benzene, 1,3,5-tris 1,1-dimethylethyl, 1,3,5-tri-t-butylbenzene, benzene, 1,3,5-tri-tert-butyl, 1,3,5-tris tert-butyl benzene, pubchem13719, intermediates-zcf02115, acmc-209cw6, 2,4,6-tri-tert-butylphenyl, ksc490m7l
PubChem CID 15089
IUPAC Name 1,3,5-tritert-butylbenzene
SMILES CC(C)(C)C1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 74°C
Color White-Yellow
Boiling Point 248°C
Quantity 5 g
Formula Weight 246.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tri-tert-butylbenzene
EINECSNumber (3)-3427
TSCA No