accessibility menu, dialog, popup

UserName

CH3H3CCH3H3CH3CCH3CH3H3CCH3

CAS RN 1460-02-2

IUPAC Name: 1,3,5-tri-tert-butylbenzene
Synonyms: 1,3,5-tri-tert-butylbenzene
Molecular Weight (g/mol): 246.44
Molecular Formula: C18H30
InChi Key: GUFMBISUSZUUCB-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)