missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri-tert-butylbenzene, 97+%

Catalog No. AAL1883209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1883209 10 g
AAL1883218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1883209 Supplier Thermo Scientific Chemicals Supplier No. L1883209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1460-02-2 | C18H30 | 246.438 g/mol

1,3,5-Tri-tert-butylbenzene is used in the preparation of sandwich complexes of scandium, yttrium and lanthanide ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Tri-tert-butylbenzene is used in the preparation of sandwich complexes of scandium, yttrium and lanthanide ions.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1460-02-2
Molecular Formula C18H30
Molecular Weight (g/mol) 246.438
MDL Number MFCD00008831
InChI Key GUFMBISUSZUUCB-UHFFFAOYSA-N
Synonym 1,3,5-tri-tert-butylbenzene, benzene, 1,3,5-tris 1,1-dimethylethyl, 1,3,5-tri-t-butylbenzene, benzene, 1,3,5-tri-tert-butyl, 1,3,5-tris tert-butyl benzene, pubchem13719, intermediates-zcf02115, acmc-209cw6, 2,4,6-tri-tert-butylphenyl, ksc490m7l
PubChem CID 15089
IUPAC Name 1,3,5-tritert-butylbenzene
SMILES CC(C)(C)C1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 69°C to 73°C
Boiling Point 121°C to 122°C (12 mmHg)
Quantity 10 g
Beilstein 1909579
Solubility Information Insoluble in water.
Formula Weight 246.44
Percent Purity ≥97%
Chemical Name or Material 1,3,5-Tri-tert-butylbenzene
EINECSNumber 215-952-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only