missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Benzenetricarboxylic acid, 98%

CAS: 554-95-0 | C9H6O6 | 210.14 g/mol

Supplier:  Thermo Scientific Chemicals A1594722

 View more versions of this product

Catalog No. AAA1594722


Only null left
Explore available promotions

Description

Description

1,3,5-Benzenetricarboxylic acid is used for the preparation of plastic, man-madefiber, water soluble alkyl resin, plasticizer and medicament intermediate. It is also used as a pharmaceutical intermediate, epoxy curing agent, plasticizers, adhesive and coating material. Further, it combines with para-hydroxypyridine to make a water-based gel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Benzenetricarboxylic acid is used for the preparation of plastic, man-madefiber, water soluble alkyl resin, plasticizer and medicament intermediate. It is also used as a pharmaceutical intermediate, epoxy curing agent, plasticizers, adhesive and coating material. Further, it combines with para-hydroxypyridine to make a water-based gel.

Solubility
Soluble in water, ethanol and methanol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

1,3,5-Benzenetricarboxylic acid
374°C to 376°C
C9H6O6
100 g
trimesic acid, 1,3,5-benzenetricarboxylic acid, trimesinic acid, trimesitinic acid, 5-carboxyisophthalic acid, 1,3,5-benzene tricarboxylic acid, 1,3,5-tricarboxybenzene, unii-ou36oo5mtn, ou36oo5mtn, chembl77562
QMKYBPDZANOJGF-UHFFFAOYSA-N
benzene-1,3,5-tricarboxylic acid
11138
210.14
554-95-0
328°C (622°F)
MFCD00002517
2053080
Soluble in water,ethanol and methanol.
OC(=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O
210.14
CHEBI:46032
98%

Chemical Identifiers

554-95-0
210.14
QMKYBPDZANOJGF-UHFFFAOYSA-N
11138
benzene-1,3,5-tricarboxylic acid
C9H6O6
MFCD00002517
trimesic acid, 1,3,5-benzenetricarboxylic acid, trimesinic acid, trimesitinic acid, 5-carboxyisophthalic acid, 1,3,5-benzene tricarboxylic acid, 1,3,5-tricarboxybenzene, unii-ou36oo5mtn, ou36oo5mtn, chembl77562
CHEBI:46032
OC(=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 209-077-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only