missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Benzenetricarboxylic acid, 98%

Catalog No. p-7022945
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1594718 50 g
AAA1594722 100 g
AAA1594730 250 g
AAA1594736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1594718 Supplier Thermo Scientific Chemicals Supplier No. A1594718

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 554-95-0 | C9H6O6 | 210.14 g/mol

1,3,5-Benzenetricarboxylic acid is used for the preparation of plastic, man-madefiber, water soluble alkyl resin, plasticizer and medicament intermediate. It is also used as a pharmaceutical intermediate, epoxy curing agent, plasticizers, adhesive and coating material. Further, it combines with para-hydroxypyridine to make a water-based gel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Benzenetricarboxylic acid is used for the preparation of plastic, man-madefiber, water soluble alkyl resin, plasticizer and medicament intermediate. It is also used as a pharmaceutical intermediate, epoxy curing agent, plasticizers, adhesive and coating material. Further, it combines with para-hydroxypyridine to make a water-based gel.

Solubility
Soluble in water, ethanol and methanol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 554-95-0
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.14
MDL Number MFCD00002517
InChI Key QMKYBPDZANOJGF-UHFFFAOYSA-N
Synonym trimesic acid, 1,3,5-benzenetricarboxylic acid, trimesinic acid, trimesitinic acid, 5-carboxyisophthalic acid, 1,3,5-benzene tricarboxylic acid, 1,3,5-tricarboxybenzene, unii-ou36oo5mtn, ou36oo5mtn, chembl77562
PubChem CID 11138
ChEBI CHEBI:46032
IUPAC Name benzene-1,3,5-tricarboxylic acid
SMILES OC(=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O

Specifications

Melting Point 374°C to 376°C
Flash Point 328°C (622°F)
Quantity 50 g
Beilstein 2053080
Solubility Information Soluble in water,ethanol and methanol.
Formula Weight 210.14
Percent Purity 98%
Chemical Name or Material 1,3,5-Benzenetricarboxylic acid
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-077-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only