accessibility menu, dialog, popup

UserName

OHOOHOOHO

CAS RN 554-95-0

IUPAC Name: benzene-1,3,5-tricarboxylic acid
Synonyms: trimesic acid trisodium salt trimesic acid trimesinic acid trimesitinic acid monosodium salt trimesic acid
Molecular Weight (g/mol): 210.14
Molecular Formula: C9H6O6
InChi Key: QMKYBPDZANOJGF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)