missing translation for 'onlineSavingsMsg'
Learn More

1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-beta-D-glucopyranose 97.0+%, TCI America™

Supplier:  TCI America T204725G

Quantity 25 g

 View more versions of this product

Catalog No. T204725G


Only null left

Description

Description

Specifications

Chemical Identifiers

10022-13-6
477.422
2728904
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)N2C(=O)C3=CC=CC=C3C2=O)OC(=O)C)OC(=O)C
C22H23NO11
DUXJAHFLYZUOPT-ACMVSEJYSA-N
[(2R,3S,4R,5R,6S)-3,4,6-triacetyloxy-5-(1,3-dioxoisoindol-2-yl)oxan-2-yl]methyl acetate

Specifications

1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-beta-D-glucopyranose
200°C
C22H23NO11
DUXJAHFLYZUOPT-ACMVSEJYSA-N
[(2R,3S,4R,5R,6S)-3,4,6-triacetyloxy-5-(1,3-dioxoisoindol-2-yl)oxan-2-yl]methyl acetate
2728904
≥97.0% (HPLC)
10022-13-6
White-Yellow
25 g
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)N2C(=O)C3=CC=CC=C3C2=O)OC(=O)C)OC(=O)C
477.422
477.42
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions